Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACTGCCTCATGGTTTTCCAGCTC[A/G]TAGAGACGAACAGGTCCCGGCTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605159 MIM: 616141 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIFM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AIFM2 - apoptosis inducing factor, mitochondria associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198696.1 | 1302 | Missense Mutation | NP_001185625.1 | |||
NM_032797.5 | 1302 | Missense Mutation | NP_116186.1 |
H2AFY2 - H2A histone family member Y2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |