Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCACCCGCTTCTACAACCGGCC[C/T]CTGCATTGTGTTTTCAATGACACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607802 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNNM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNNM1 - cyclin and CBS domain divalent metal cation transport mediator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020348.2 | 1804 | Silent Mutation | CCC,CCT | P,P 505 | NP_065081.2 | |
XM_005269692.3 | 1804 | Silent Mutation | CCC,CCT | P,P 505 | XP_005269749.2 | |
XM_011539629.1 | 1804 | Silent Mutation | CCC,CCT | P,P 505 | XP_011537931.1 | |
XM_011539630.1 | 1804 | Silent Mutation | CCC,CCT | P,P 505 | XP_011537932.1 | |
XM_011539631.2 | 1804 | Silent Mutation | CCC,CCT | P,P 505 | XP_011537933.1 |