Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCTGGACTTCTCCTCTGGCTCC[A/G]GCTTCGCTGTAGCCTCCAGCAGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611207 | ||||||||||||||||||||
Literature Links: |
DNAJC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJC1 - DnaJ heat shock protein family (Hsp40) member C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022365.3 | 1657 | Missense Mutation | CCG,CTG | P,L 457 | NP_071760.2 | |
XM_011519614.2 | 1657 | Intron | XP_011517916.1 | |||
XM_017016536.1 | 1657 | Intron | XP_016872025.1 |