Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGCATCCGCCTCTTCAATCTTAC[C/T]TCCACCTTCAGTCGCCACTCGGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608842 MIM: 616932 MIM: 607185 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHCHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHCHD1 - coiled-coil-helix-coiled-coil-helix domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FUT11 - fucosyltransferase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284194.1 | 663 | Silent Mutation | ACC,ACT | T,T 168 | NP_001271123.1 | |
NM_173540.2 | 663 | Silent Mutation | ACC,ACT | T,T 168 | NP_775811.2 | |
XM_006717656.3 | 663 | Silent Mutation | ACC,ACT | T,T 168 | XP_006717719.1 |
SEC24C - SEC24 homolog C, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004922.3 | 663 | Intron | NP_004913.2 | |||
NM_198597.2 | 663 | Intron | NP_940999.1 | |||
XM_011540380.2 | 663 | Intron | XP_011538682.1 | |||
XM_011540381.2 | 663 | Intron | XP_011538683.1 | |||
XM_017016967.1 | 663 | Intron | XP_016872456.1 | |||
XM_017016968.1 | 663 | Intron | XP_016872457.1 |