Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATCTGTCATTCTCAGGTAACAG[C/G]GACACCTAGACAATTAAAAATTAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607472 | ||||||||||||||||||||
Literature Links: |
YME1L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
YME1L1 - YME1 like 1 ATPase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253866.1 | 2389 | Silent Mutation | TCC,TCG | S,S 542 | NP_001240795.1 | |
NM_014263.3 | 2389 | Silent Mutation | TCC,TCG | S,S 575 | NP_055078.1 | |
NM_139312.2 | 2389 | Silent Mutation | TCC,TCG | S,S 632 | NP_647473.1 | |
XM_011519300.2 | 2389 | Silent Mutation | TCC,TCG | S,S 599 | XP_011517602.1 |