Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCATAAAGCTAAGGTTAAAATGG[C/T]GAGAACCAACAATCCAAGAAGTATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604063 | ||||||||||||||||||||
Literature Links: |
ITGA8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITGA8 - integrin subunit alpha 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291494.1 | 3598 | Missense Mutation | ACC,GCC | T,A 1012 | NP_001278423.1 | |
NM_003638.2 | 3598 | Missense Mutation | ACC,GCC | T,A 1027 | NP_003629.2 | |
XM_011519752.2 | 3598 | Intron | XP_011518054.1 |