Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCAGGTCTTGGGAACAGGTGTCC[C/T]TCTGCTATAGAGGTAAATATGATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POLA2 - DNA polymerase alpha 2, accessory subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002689.3 | Intron | NP_002680.2 | ||||
XM_011544877.2 | Intron | XP_011543179.1 | ||||
XM_011544878.2 | Intron | XP_011543180.1 | ||||
XM_011544879.1 | Intron | XP_011543181.1 | ||||
XM_011544881.2 | Intron | XP_011543183.1 | ||||
XM_017017452.1 | Intron | XP_016872941.1 |