Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCCTCCCAGCAAGCACTGACTC[A/G]TCGCAGACTTTTAACTAGTCCAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616225 MIM: 601644 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATG2A - autophagy related 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP2R5B - protein phosphatase 2 regulatory subunit B'beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006244.3 | 774 | Intron | NP_006235.1 | |||
XM_011545132.2 | 774 | Intron | XP_011543434.1 | |||
XM_011545133.1 | 774 | Intron | XP_011543435.1 | |||
XM_011545134.2 | 774 | UTR 5 | XP_011543436.1 |