Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCTGCAACATGGATAATAAGGA[C/T]TTGGTGTGGCTTGAGGAAATCCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604411 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INCENP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INCENP - inner centromere protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040694.1 | 254 | Silent Mutation | GAC,GAT | D,D 30 | NP_001035784.1 | |
NM_020238.2 | 254 | Silent Mutation | GAC,GAT | D,D 30 | NP_064623.2 | |
XM_006718533.2 | 254 | Silent Mutation | GAC,GAT | D,D 30 | XP_006718596.1 | |
XM_011544995.2 | 254 | Silent Mutation | GAC,GAT | D,D 30 | XP_011543297.1 | |
XM_011544996.2 | 254 | Silent Mutation | GAC,GAT | D,D 30 | XP_011543298.1 | |
XM_011544997.2 | 254 | Silent Mutation | GAC,GAT | D,D 30 | XP_011543299.1 | |
XM_011544998.2 | 254 | Silent Mutation | GAC,GAT | D,D 30 | XP_011543300.1 |