Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAACTCTCTGGTTTTTGCAGCCTT[C/T]TCCACTCGTCTGTTTGCTGTCCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601134 | ||||||||||||||||||||
Literature Links: |
STT3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STT3A - STT3A, catalytic subunit of the oligosaccharyltransferase complex | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278503.1 | 252 | Silent Mutation | TTC,TTT | F,F 31 | NP_001265432.1 | |
NM_001278504.1 | 252 | UTR 5 | NP_001265433.1 | |||
NM_152713.4 | 252 | Silent Mutation | TTC,TTT | F,F 31 | NP_689926.1 | |
XM_011542807.2 | 252 | Silent Mutation | TTC,TTT | F,F 31 | XP_011541109.1 |
STT3A-AS1 - STT3A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |