Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAGAGGCGGGCAGCCTCGGCGAA[G/T]CCCACCCTGTTGTTGTCACGGTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116840 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTSD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTSD - cathepsin D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001909.4 | 1348 | Silent Mutation | GGA,GGC | G,G 405 | NP_001900.1 |
IFITM10 - interferon induced transmembrane protein 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |