Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATGGAGAGGCAGAGGAAGAGGT[A/G]GCGGAGGGTGGGGCAGCGCAGCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607582 | ||||||||||||||||||||
Literature Links: |
SLC22A6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC22A6 - solute carrier family 22 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004790.4 | 1342 | Missense Mutation | CAC,TAC | H,Y 337 | NP_004781.2 | |
NM_153276.2 | 1342 | Missense Mutation | CAC,TAC | H,Y 337 | NP_695008.1 | |
NM_153277.2 | 1342 | Missense Mutation | CAC,TAC | H,Y 337 | NP_695009.1 | |
NM_153278.2 | 1342 | Missense Mutation | CAC,TAC | H,Y 337 | NP_695010.1 | |
XM_017018562.1 | 1342 | Missense Mutation | CAC,TAC | H,Y 338 | XP_016874051.1 |