Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCACACTATGTCAGAGTTGATGA[A/G]GAGGTATCTGTCACCTACACAGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180385 | ||||||||||||||||||||
Literature Links: |
LMO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LMO2 - LIM domain only 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142315.1 | 1200 | Missense Mutation | CTC,TTC | L,F 137 | NP_001135787.1 | |
NM_001142316.1 | 1200 | Missense Mutation | CTC,TTC | L,F 137 | NP_001135788.1 | |
NM_005574.3 | 1200 | Missense Mutation | CTC,TTC | L,F 206 | NP_005565.2 | |
XM_005252921.2 | 1200 | Missense Mutation | CTC,TTC | L,F 137 | XP_005252978.1 | |
XM_017017727.1 | 1200 | Missense Mutation | CTC,TTC | L,F 137 | XP_016873216.1 | |
XM_017017728.1 | 1200 | Missense Mutation | CTC,TTC | L,F 137 | XP_016873217.1 | |
XM_017017729.1 | 1200 | Missense Mutation | CTC,TTC | L,F 137 | XP_016873218.1 | |
XM_017017730.1 | 1200 | Missense Mutation | CTC,TTC | L,F 137 | XP_016873219.1 | |
XM_017017731.1 | 1200 | Missense Mutation | CTC,TTC | L,F 137 | XP_016873220.1 | |
XM_017017732.1 | 1200 | Missense Mutation | CTC,TTC | L,F 137 | XP_016873221.1 | |
XM_017017733.1 | 1200 | Missense Mutation | CTC,TTC | L,F 137 | XP_016873222.1 |