Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGAACCGCCAACAGCGCCCCGG[C/G]ACCATCAAGCATGGATCGGCGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609201 | ||||||||||||||||||||
Literature Links: |
UBASH3B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBASH3B - ubiquitin associated and SH3 domain containing B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032873.4 | 459 | Silent Mutation | GGC,GGG | G,G 33 | NP_116262.2 | |
XM_005271712.3 | 459 | Intron | XP_005271769.1 | |||
XM_011543041.2 | 459 | Intron | XP_011541343.1 | |||
XM_011543042.1 | 459 | UTR 5 | XP_011541344.1 |