Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTAGCCTTGTAAACTCAGCCTT[A/C]CAGCTCTCAATCATTATAATTCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD42 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD42 - ankyrin repeat domain 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300972.1 | Intron | NP_001287901.1 | ||||
NM_001300973.1 | Intron | NP_001287902.1 | ||||
NM_001300975.1 | Intron | NP_001287904.1 | ||||
NM_001300976.1 | Intron | NP_001287905.1 | ||||
NM_001300977.1 | Intron | NP_001287906.1 | ||||
NM_182603.3 | Intron | NP_872409.2 |
LOC100506282 - uncharacterized LOC100506282 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCF11-AS1 - PCF11 anstisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |