Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTCGCGCTGTCTCCCCAGGGGGA[A/G]ATAATCGAGGGCTGCCGCCTACCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601409 MIM: 610330 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KAT5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KAT5 - lysine acetyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206833.1 | 284 | Silent Mutation | GAA,GAG | E,E 39 | NP_001193762.1 | |
NM_006388.3 | 284 | Silent Mutation | GAA,GAG | E,E 6 | NP_006379.2 | |
NM_182709.2 | 284 | Silent Mutation | GAA,GAG | E,E 6 | NP_874368.1 | |
NM_182710.2 | 284 | Silent Mutation | GAA,GAG | E,E 39 | NP_874369.1 | |
XM_006718421.2 | 284 | Silent Mutation | GAA,GAG | E,E 6 | XP_006718484.1 |
RNASEH2C - ribonuclease H2 subunit C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |