Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCATCATCTTCATTATAGGTGT[C/G]AAATTCATCATCACCATCAAAACTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179616 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf74 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf74 - chromosome 11 open reading frame 74 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAG2 - recombination activating gene 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000536.3 | 1403 | Missense Mutation | CAC,GAC | H,D 400 | NP_000527.2 | |
NM_001243785.1 | 1403 | Missense Mutation | CAC,GAC | H,D 400 | NP_001230714.1 | |
NM_001243786.1 | 1403 | Missense Mutation | CAC,GAC | H,D 400 | NP_001230715.1 |