Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCAATTGTATCTATTGCTTGGCA[A/G]AAAGAAGTTTCAGGGCACATTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 158070 MIM: 603222 MIM: 603223 MIM: 603224 MIM: 603225 MIM: 603226 MIM: 603227 MIM: 603228 MIM: 603229 MIM: 603230 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC3A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC3A2 - solute carrier family 3 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNHG1 - small nucleolar RNA host gene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD22 - small nucleolar RNA, C/D box 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD25 - small nucleolar RNA, C/D box 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD26 - small nucleolar RNA, C/D box 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD27 - small nucleolar RNA, C/D box 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD28 - small nucleolar RNA, C/D box 28 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD29 - small nucleolar RNA, C/D box 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD30 - small nucleolar RNA, C/D box 30 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD31 - small nucleolar RNA, C/D box 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |