Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGGGGTCCCCCAGGCCCTCTAC[C/T]CACCATGTTCAGTCCCTCTGTCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600466 MIM: 608204 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH3B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH3B1 - aldehyde dehydrogenase 3 family member B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000694.3 | Intron | NP_000685.1 | ||||
NM_001030010.2 | Intron | NP_001025181.1 | ||||
NM_001161473.2 | Intron | NP_001154945.1 | ||||
NM_001290058.1 | Intron | NP_001276987.1 | ||||
NM_001290059.1 | Intron | NP_001276988.1 |
UNC93B1 - unc-93 homolog B1 (C. elegans) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |