Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCCTCGAGTGGGCTCAGGAGAA[A/G]CGTAAGCTGAGCGTGTTGCATATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616587 MIM: 182180 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM118B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM118B - family with sequence similarity 118 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024556.3 | 705 | Silent Mutation | AAA,AAG | K,K 197 | NP_078832.1 | |
XM_005271665.3 | 705 | Silent Mutation | AAA,AAG | K,K 197 | XP_005271722.1 | |
XM_011542977.2 | 705 | Silent Mutation | AAA,AAG | K,K 197 | XP_011541279.1 | |
XM_011542978.2 | 705 | Silent Mutation | AAA,AAG | K,K 42 | XP_011541280.1 | |
XM_017018285.1 | 705 | Silent Mutation | AAA,AAG | K,K 197 | XP_016873774.1 | |
XM_017018286.1 | 705 | Silent Mutation | AAA,AAG | K,K 42 | XP_016873775.1 |
SRPRA - SRP receptor alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177842.1 | 705 | Intron | NP_001171313.1 | |||
NM_003139.3 | 705 | Intron | NP_003130.2 | |||
XM_017018179.1 | 705 | Intron | XP_016873668.1 |