Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGAGGAACCCGTGTCTGTGCAC[A/G]GCAGCACGGTCTTCAAGGAGCTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605242 | ||||||||||||||||||||
Literature Links: |
USH1C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USH1C - USH1 protein network component harmonin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297764.1 | 2725 | Intron | NP_001284693.1 | |||
NM_005709.3 | 2725 | Intron | NP_005700.2 | |||
NM_153676.3 | 2725 | Silent Mutation | GCC,GCT | A,A 872 | NP_710142.1 | |
XM_011519832.2 | 2725 | Intron | XP_011518134.1 | |||
XM_011519834.2 | 2725 | Intron | XP_011518136.1 | |||
XM_017017072.1 | 2725 | Intron | XP_016872561.1 | |||
XM_017017073.1 | 2725 | Intron | XP_016872562.1 | |||
XM_017017074.1 | 2725 | Intron | XP_016872563.1 | |||
XM_017017075.1 | 2725 | Silent Mutation | GCC,GCT | A,A 872 | XP_016872564.1 |