Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGGGGATGAAGACAGCCTCCGGG[A/G]ACTACATCGACTCGTCATGGGAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607901 MIM: 605063 | ||||||||||||||||||||
Literature Links: |
FERMT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FERMT3 - fermitin family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031471.5 | 177 | Missense Mutation | AAC,GAC | N,D 10 | NP_113659.3 | |
NM_178443.2 | 177 | Missense Mutation | AAC,GAC | N,D 10 | NP_848537.1 | |
XM_011545294.2 | 177 | Missense Mutation | AAC,GAC | N,D 10 | XP_011543596.1 | |
XM_011545295.2 | 177 | Intron | XP_011543597.1 | |||
XM_017018398.1 | 177 | Missense Mutation | AAC,GAC | N,D 10 | XP_016873887.1 | |
XM_017018399.1 | 177 | Intron | XP_016873888.1 |
STIP1 - stress induced phosphoprotein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |