Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATCTCAAGATAGAGATTTTGAA[A/G]CTATGGCTAAATTACATATTCCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611271 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF18A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF18A - kinesin family member 18A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL15 - methyltransferase like 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113528.1 | 654 | Missense Mutation | ACT,GCT | T,A 67 | NP_001107000.1 | |
NM_001297775.1 | 654 | Missense Mutation | ACT,GCT | T,A 67 | NP_001284704.1 | |
NM_152636.2 | 654 | Missense Mutation | ACT,GCT | T,A 67 | NP_689849.2 | |
XM_011519935.2 | 654 | UTR 5 | XP_011518237.1 | |||
XM_011519937.2 | 654 | Intron | XP_011518239.1 | |||
XM_011519940.2 | 654 | Missense Mutation | ACT,GCT | T,A 67 | XP_011518242.1 | |
XM_011519941.2 | 654 | Missense Mutation | ACT,GCT | T,A 67 | XP_011518243.1 | |
XM_017017297.1 | 654 | Missense Mutation | ACT,GCT | T,A 67 | XP_016872786.1 | |
XM_017017298.1 | 654 | UTR 5 | XP_016872787.1 | |||
XM_017017299.1 | 654 | Intron | XP_016872788.1 | |||
XM_017017300.1 | 654 | UTR 5 | XP_016872789.1 | |||
XM_017017301.1 | 654 | Missense Mutation | ACT,GCT | T,A 67 | XP_016872790.1 | |
XM_017017302.1 | 654 | Intron | XP_016872791.1 | |||
XM_017017303.1 | 654 | UTR 5 | XP_016872792.1 |