Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCACCACCCACTCCACGTCGTC[A/G]ATGCCCTCGCCGGCCCCCTCGCCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613991 MIM: 605888 | ||||||||||||||||||||
Literature Links: |
CDC42BPG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC42BPG - CDC42 binding protein kinase gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EHD1 - EH domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282444.1 | 1427 | Silent Mutation | ATC,ATT | I,I 434 | NP_001269373.1 | |
NM_001282445.1 | 1427 | Silent Mutation | ATC,ATT | I,I 448 | NP_001269374.1 | |
NM_006795.3 | 1427 | Silent Mutation | ATC,ATT | I,I 434 | NP_006786.2 | |
XM_011544739.1 | 1427 | Silent Mutation | ATC,ATT | I,I 434 | XP_011543041.1 |