Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTAATCATACCTTGTTAATTTCC[A/G]TTTCCTTTTGGCAGCAGTAGTCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611271 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF18A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF18A - kinesin family member 18A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031217.3 | 1381 | Missense Mutation | CGG,TGG | R,W 831 | NP_112494.3 | |
XM_017018379.1 | 1381 | Missense Mutation | CGG,TGG | R,W 831 | XP_016873868.1 | |
XM_017018380.1 | 1381 | Missense Mutation | CGG,TGG | R,W 387 | XP_016873869.1 |