Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTTCAACAGTCCTTGTTGGGGC[A/G]TGATGGTTTTGCCGATGGTGCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608817 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC4C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC4C - leucine rich repeat containing 4C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258419.1 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | NP_001245348.1 | |
NM_020929.2 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | NP_065980.1 | |
XM_011520238.2 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_011518540.1 | |
XM_011520239.2 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_011518541.1 | |
XM_011520240.2 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_011518542.1 | |
XM_011520241.2 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_011518543.1 | |
XM_011520242.2 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_011518544.1 | |
XM_011520243.2 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_011518545.1 | |
XM_011520244.2 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_011518546.1 | |
XM_017018070.1 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_016873559.1 | |
XM_017018071.1 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_016873560.1 | |
XM_017018072.1 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_016873561.1 | |
XM_017018073.1 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_016873562.1 | |
XM_017018074.1 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_016873563.1 | |
XM_017018075.1 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_016873564.1 | |
XM_017018076.1 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_016873565.1 | |
XM_017018077.1 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_016873566.1 | |
XM_017018078.1 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_016873567.1 | |
XM_017018079.1 | 2904 | Silent Mutation | CAC,CAT | H,H 560 | XP_016873568.1 |