Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCCCCGGACCCCAGCTCTGGCA[C/G]TTGCTTCAGGCCCTTGTGTCTATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 209901 MIM: 606818 | ||||||||||||||||||||
Literature Links: |
BBS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BBS1 - Bardet-Biedl syndrome 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024649.4 | 328 | Missense Mutation | CTT,GTT | L,V 106 | NP_078925.3 |
DPP3 - dipeptidyl peptidase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZDHHC24 - zinc finger DHHC-type containing 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |