Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCACGCGCAGCAGGTTGGCTACA[C/T]GTTTGGCCATGTCTAGAGAGAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606372 MIM: 601021 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRNA10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRNA10 - cholinergic receptor nicotinic alpha 10 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP98 - nucleoporin 98 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005387.6 | 5397 | Intron | NP_005378.4 | |||
NM_016320.4 | 5397 | Missense Mutation | CAT,CGT | H,R 1733 | NP_057404.2 | |
NM_139131.4 | 5397 | Intron | NP_624357.1 | |||
NM_139132.3 | 5397 | Missense Mutation | CAT,CGT | H,R 1659 | NP_624358.2 |