Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGGTCCGACGGTGTGGCCAAGA[C/T]GAGCCCCAAGGACCTGTTTGGTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614988 | ||||||||||||||||||||
Literature Links: |
EPS8L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPS8L2 - EPS8 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022772.3 | 327 | Missense Mutation | ACG,ATG | T,M 27 | NP_073609.2 | |
XM_017018130.1 | 327 | Missense Mutation | ACG,ATG | T,M 27 | XP_016873619.1 | |
XM_017018131.1 | 327 | Missense Mutation | ACG,ATG | T,M 27 | XP_016873620.1 | |
XM_017018132.1 | 327 | Missense Mutation | ACG,ATG | T,M 27 | XP_016873621.1 |
TMEM80 - transmembrane protein 80 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |