Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTATCAAAAACGTCAGTATAAAT[C/T]GGGTGCCGTGCTATGCACCTGTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602493 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UVRAG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UVRAG - UV radiation resistance associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003369.3 | Intron | NP_003360.2 | ||||
XM_011545226.2 | Intron | XP_011543528.1 | ||||
XM_011545227.2 | Intron | XP_011543529.1 | ||||
XM_011545228.2 | Intron | XP_011543530.1 | ||||
XM_017018224.1 | Intron | XP_016873713.1 | ||||
XM_017018225.1 | Intron | XP_016873714.1 | ||||
XM_017018226.1 | Intron | XP_016873715.1 | ||||
XM_017018227.1 | Intron | XP_016873716.1 |