Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAGAGAAGTTCTTGAAGGGAGAA[A/C]CCAAAGTCCTTGGGGTAAGTTGCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606547 | ||||||||||||||||||||
Literature Links: |
MS4A4A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MS4A4A - membrane spanning 4-domains A4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243266.1 | Intron | NP_001230195.1 | ||||
NM_024021.3 | Intron | NP_076926.2 | ||||
NM_148975.2 | Intron | NP_683876.1 | ||||
XM_017017909.1 | Intron | XP_016873398.1 |