Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATACATTCTGAAGACCGCCGCAC[A/T]CGCATCCATCGAGGCACGATTGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606374 MIM: 104160 | ||||||||||||||||||||
Literature Links: |
B3GAT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
B3GAT3 - beta-1,3-glucuronyltransferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GANAB - glucosidase II alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278192.1 | 2638 | Silent Mutation | CGA,CGT | R,R 723 | NP_001265121.1 | |
NM_001278193.1 | 2638 | Silent Mutation | CGA,CGT | R,R 701 | NP_001265122.1 | |
NM_001278194.1 | 2638 | Silent Mutation | CGA,CGT | R,R 718 | NP_001265123.1 | |
NM_198334.2 | 2638 | Silent Mutation | CGA,CGT | R,R 815 | NP_938148.1 | |
NM_198335.3 | 2638 | Silent Mutation | CGA,CGT | R,R 837 | NP_938149.2 | |
XM_017017412.1 | 2638 | Silent Mutation | CGA,CGT | R,R 718 | XP_016872901.1 | |
XM_017017413.1 | 2638 | Silent Mutation | CGA,CGT | R,R 574 | XP_016872902.1 | |
XM_017017414.1 | 2638 | Silent Mutation | CGA,CGT | R,R 574 | XP_016872903.1 |