Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGCTGTGGTGGTGGCAATGCGC[A/G]GAGAACGGCGGGTTCCACTGCGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604113 MIM: 164009 | ||||||||||||||||||||
Literature Links: |
IL18BP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL18BP - interleukin 18 binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUMA1 - nuclear mitotic apparatus protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286561.1 | 6468 | Missense Mutation | CCG,CTG | P,L 2074 | NP_001273490.1 | |
NM_006185.3 | 6468 | Missense Mutation | CCG,CTG | P,L 2088 | NP_006176.2 | |
XM_006718564.1 | 6468 | Missense Mutation | CCG,CTG | P,L 2088 | XP_006718627.1 | |
XM_011545054.1 | 6468 | Missense Mutation | CCG,CTG | P,L 2106 | XP_011543356.1 | |
XM_011545055.1 | 6468 | Missense Mutation | CCG,CTG | P,L 2106 | XP_011543357.1 | |
XM_011545056.2 | 6468 | Missense Mutation | CCG,CTG | P,L 2106 | XP_011543358.1 | |
XM_011545057.2 | 6468 | Missense Mutation | CCG,CTG | P,L 2106 | XP_011543359.1 | |
XM_011545058.1 | 6468 | Missense Mutation | CCG,CTG | P,L 2106 | XP_011543360.1 | |
XM_011545059.2 | 6468 | Missense Mutation | CCG,CTG | P,L 2106 | XP_011543361.1 | |
XM_011545060.2 | 6468 | Missense Mutation | CCG,CTG | P,L 2106 | XP_011543362.1 | |
XM_011545061.2 | 6468 | Missense Mutation | CCG,CTG | P,L 2106 | XP_011543363.1 | |
XM_011545062.2 | 6468 | Missense Mutation | CCG,CTG | P,L 2106 | XP_011543364.1 | |
XM_011545063.2 | 6468 | Missense Mutation | CCG,CTG | P,L 2106 | XP_011543365.1 | |
XM_011545064.1 | 6468 | Missense Mutation | CCG,CTG | P,L 2106 | XP_011543366.1 | |
XM_011545065.1 | 6468 | Missense Mutation | CCG,CTG | P,L 2092 | XP_011543367.1 | |
XM_011545066.1 | 6468 | Missense Mutation | CCG,CTG | P,L 2074 | XP_011543368.1 | |
XM_017017830.1 | 6468 | Missense Mutation | CCG,CTG | P,L 2074 | XP_016873319.1 | |
XM_017017831.1 | 6468 | Missense Mutation | CCG,CTG | P,L 2074 | XP_016873320.1 |
RNF121 - ring finger protein 121 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |