Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTTAGAGTTTAAAAGAAACTTTG[A/G]GACTAATTCAATTTCTTCATTTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612803 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928865 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101928865 - uncharacterized LOC101928865 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NARS2 - asparaginyl-tRNA synthetase 2, mitochondrial (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243251.1 | 2052 | UTR 3 | NP_001230180.1 | |||
NM_024678.5 | 2052 | UTR 3 | NP_078954.4 | |||
XM_011545253.1 | 2052 | UTR 3 | XP_011543555.1 | |||
XM_017018302.1 | 2052 | Intron | XP_016873791.1 | |||
XM_017018303.1 | 2052 | UTR 3 | XP_016873792.1 | |||
XM_017018304.1 | 2052 | UTR 3 | XP_016873793.1 |