Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAAGCTCTATGCTAGGTAAGGCT[G/T]GTATCCAAGCTGGCTGCCGAGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605473 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OLFM5P PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OLFM5P - olfactomedin family member 5, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR51B5 - olfactory receptor family 51 subfamily B member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBQLN3 - ubiquilin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBQLNL - ubiquilin-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145053.4 | 1427 | Missense Mutation | CAA,CCA | Q,P 381 | NP_659490.4 |