Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGTGATGATCCCCAGCCTTCTG[C/T]GGGACGTGCAGCTGAGTGGGCCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603845 MIM: 601926 | ||||||||||||||||||||
Literature Links: |
NDUFC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDUFC2 - NADH:ubiquinone oxidoreductase subunit C2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFC2-KCTD14 - NDUFC2-KCTD14 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001203260.1 | 133 | Intron | NP_001190189.1 | |||
NM_001203261.1 | 133 | Intron | NP_001190190.1 | |||
NM_001203262.1 | 133 | Intron | NP_001190191.1 |
THRSP - thyroid hormone responsive | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003251.3 | 133 | Missense Mutation | CGG,TGG | R,W 38 | NP_003242.1 |