Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAAGACTCACCTTCTCACCCCA[A/G]TCAGTGCCCCAGCTGTTCTTGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102574 MIM: 603539 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTN3 - actinin alpha 3 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104.3 | 1208 | Intron | NP_001095.2 | |||
NM_001258371.2 | 1208 | Intron | NP_001245300.2 |
CTSF - cathepsin F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003793.3 | 1208 | Silent Mutation | GAC,GAT | D,D 456 | NP_003784.2 | |
XM_011545328.2 | 1208 | Silent Mutation | GAC,GAT | D,D 396 | XP_011543630.1 |