Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGAGACCCAATTGCAGGGACTG[G/T]AGTCTGCATCTGGATGAGCTGGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601814 MIM: 606683 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FXYD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FXYD2 - FXYD domain containing ion transport regulator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FXYD6 - FXYD domain containing ion transport regulator 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164831.2 | 1719 | UTR 3 | NP_001158303.1 | |||
NM_001164832.2 | 1719 | UTR 3 | NP_001158304.1 | |||
NM_001164836.2 | 1719 | UTR 3 | NP_001158308.1 | |||
NM_001164837.2 | 1719 | UTR 3 | NP_001158309.1 | |||
NM_022003.3 | 1719 | UTR 3 | NP_071286.1 |
FXYD6-FXYD2 - FXYD6-FXYD2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204268.1 | 1719 | Intron | NP_001191197.1 | |||
NM_001243598.2 | 1719 | Intron | NP_001230527.1 |