Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGCTGCCTCTCCTGCCTGCGCT[A/G]GTCCTCAGCACCAGGTGAGTCCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608178 | ||||||||||||||||||||
Literature Links: |
LUZP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LUZP2 - leucine zipper protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009909.3 | 322 | Silent Mutation | CTA,CTG | L,L 16 | NP_001009909.2 | |
NM_001252008.1 | 322 | UTR 5 | NP_001238937.1 | |||
NM_001252010.1 | 322 | Silent Mutation | CTA,CTG | L,L 16 | NP_001238939.1 | |
XM_011520056.2 | 322 | Missense Mutation | AGT,GGT | S,G 144 | XP_011518358.1 | |
XM_017017648.1 | 322 | UTR 5 | XP_016873137.1 | |||
XM_017017649.1 | 322 | UTR 5 | XP_016873138.1 | |||
XM_017017650.1 | 322 | Silent Mutation | CTA,CTG | L,L 16 | XP_016873139.1 | |
XM_017017651.1 | 322 | Silent Mutation | CTA,CTG | L,L 16 | XP_016873140.1 |