Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGTCAGGCCCTCAAGCTGGTGG[C/T]TGAGCTTCTCCTGCGTGTCCTGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107690 MIM: 107720 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOA4 - apolipoprotein A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000482.3 | 808 | Missense Mutation | AAC,AGC | N,S 235 | NP_000473.2 |
APOC3 - apolipoprotein C3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |