Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTGCTCCCGTTCATACCGGGCCA[C/T]GTCATACAGAGAATTCCGTGTAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602571 | ||||||||||||||||||||
Literature Links: |
RBM4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RBM4 - RNA binding motif protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198843.1 | 1160 | Intron | NP_001185772.1 | |||
NM_001198844.1 | 1160 | Intron | NP_001185773.1 | |||
NM_002896.3 | 1160 | Intron | NP_002887.2 |
RBM4B - RNA binding motif protein 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286135.1 | 1160 | Intron | NP_001273064.1 | |||
NM_031492.3 | 1160 | Missense Mutation | ATG,GTG | M,V 342 | NP_113680.1 | |
XM_011545297.2 | 1160 | Missense Mutation | ATG,GTG | M,V 342 | XP_011543599.1 | |
XM_017018402.1 | 1160 | Missense Mutation | ATG,GTG | M,V 342 | XP_016873891.1 |