Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCCCAGCACGTCCATTCTTCCCA[A/G]ACAGCTCATGACTCCATGGTATGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137207 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC32 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC32 - leucine rich repeat containing 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128922.1 | 2399 | UTR 3 | NP_001122394.1 | |||
NM_005512.2 | 2399 | UTR 3 | NP_005503.1 | |||
XM_005273902.3 | 2399 | UTR 3 | XP_005273959.1 | |||
XM_005273903.4 | 2399 | UTR 3 | XP_005273960.2 | |||
XM_011544935.2 | 2399 | UTR 3 | XP_011543237.1 | |||
XM_017017527.1 | 2399 | UTR 3 | XP_016873016.1 |