Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCCCTCCCTCCCAGAATAGCTC[A/T]CAGTTCATGAAGCACACCAAACGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602013 MIM: 602946 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLR2G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POLR2G - RNA polymerase II subunit G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAF6L - TATA-box binding protein associated factor 6 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006473.3 | 249 | Silent Mutation | TCA,TCT | S,S 52 | NP_006464.1 | |
XM_005273714.1 | 249 | Silent Mutation | TCA,TCT | S,S 52 | XP_005273771.1 | |
XM_017017100.1 | 249 | Intron | XP_016872589.1 |