Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCTTCTTCACCTTCTTGACCTC[G/T]CCCGTCTTGCCGTAGTTGCGCAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600958 MIM: 165170 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYBPC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYBPC3 - myosin binding protein C, cardiac | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPI1 - Spi-1 proto-oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080547.1 | 2388 | Silent Mutation | GGA,GGC | G,G 240 | NP_001074016.1 | |
NM_003120.2 | 2388 | Silent Mutation | GGA,GGC | G,G 239 | NP_003111.2 | |
XM_011520307.1 | 2388 | Silent Mutation | GGA,GGC | G,G 152 | XP_011518609.1 | |
XM_017018173.1 | 2388 | Silent Mutation | GGA,GGC | G,G 152 | XP_016873662.1 |