Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTGTGCCTGGCGTGAAGTCTCA[A/G]TGCTGCCCCATCCCACTGCTGCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611947 MIM: 607146 | ||||||||||||||||||||
Literature Links: |
CCDC153 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC153 - coiled-coil domain containing 153 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145018.1 | 849 | Silent Mutation | CTG,TTG | L,L 182 | NP_001138490.1 | |
XM_006718818.3 | 849 | Silent Mutation | CTG,TTG | L,L 154 | XP_006718881.1 | |
XM_011542763.2 | 849 | Silent Mutation | CTG,TTG | L,L 342 | XP_011541065.1 | |
XM_011542764.2 | 849 | Silent Mutation | CTG,TTG | L,L 342 | XP_011541066.1 | |
XM_011542770.2 | 849 | Silent Mutation | CTG,TTG | L,L 182 | XP_011541072.1 | |
XM_011542771.2 | 849 | Silent Mutation | CTG,TTG | L,L 174 | XP_011541073.1 | |
XM_011542772.2 | 849 | Silent Mutation | CTG,TTG | L,L 174 | XP_011541074.1 | |
XM_011542774.2 | 849 | Silent Mutation | CTG,TTG | L,L 244 | XP_011541076.1 | |
XM_017017577.1 | 849 | Silent Mutation | CTG,TTG | L,L 270 | XP_016873066.1 | |
XM_017017578.1 | 849 | Intron | XP_016873067.1 | |||
XM_017017579.1 | 849 | Silent Mutation | CTG,TTG | L,L 111 | XP_016873068.1 | |
XM_017017580.1 | 849 | Silent Mutation | CTG,TTG | L,L 182 | XP_016873069.1 |
NLRX1 - NLR family member X1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDZD3 - PDZ domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168468.1 | 849 | Intron | NP_001161940.1 | |||
NM_024791.3 | 849 | Intron | NP_079067.3 | |||
XM_011543000.2 | 849 | Intron | XP_011541302.1 | |||
XM_011543001.2 | 849 | Intron | XP_011541303.1 | |||
XM_011543002.2 | 849 | Intron | XP_011541304.1 | |||
XM_011543005.2 | 849 | Intron | XP_011541307.1 | |||
XM_017018334.1 | 849 | Intron | XP_016873823.1 | |||
XM_017018335.1 | 849 | Intron | XP_016873824.1 | |||
XM_017018336.1 | 849 | Intron | XP_016873825.1 | |||
XM_017018337.1 | 849 | Intron | XP_016873826.1 |