Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCATTCTCAGTAACTCGTTGGGT[A/G]TAAAACAAGATATAAGCTTGAGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601870 MIM: 610993 | ||||||||||||||||||||
Literature Links: |
METAP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
METAP2 - methionyl aminopeptidase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP44 - ubiquitin specific peptidase 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042403.2 | 2303 | Silent Mutation | NP_001035862.1 | |||
NM_001278393.1 | 2303 | Silent Mutation | NP_001265322.1 | |||
NM_032147.4 | 2303 | Silent Mutation | NP_115523.2 | |||
XM_005269172.1 | 2303 | Silent Mutation | XP_005269229.1 | |||
XM_005269173.1 | 2303 | Silent Mutation | XP_005269230.1 | |||
XM_005269174.1 | 2303 | Silent Mutation | XP_005269231.1 | |||
XM_011538800.2 | 2303 | Silent Mutation | XP_011537102.1 | |||
XM_011538801.1 | 2303 | Nonsense Mutation | XP_011537103.1 | |||
XM_011538802.1 | 2303 | Silent Mutation | XP_011537104.1 | |||
XM_011538803.1 | 2303 | Silent Mutation | XP_011537105.1 | |||
XM_011538804.2 | 2303 | Silent Mutation | XP_011537106.1 | |||
XM_011538805.1 | 2303 | Silent Mutation | XP_011537107.1 | |||
XM_011538806.2 | 2303 | Intron | XP_011537108.1 | |||
XM_017020009.1 | 2303 | Silent Mutation | XP_016875498.1 | |||
XM_017020010.1 | 2303 | Silent Mutation | XP_016875499.1 | |||
XM_017020011.1 | 2303 | Silent Mutation | XP_016875500.1 | |||
XM_017020012.1 | 2303 | Intron | XP_016875501.1 |