Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTTCGCCTCCACTGACTCCCGC[C/T]GCACCAGCCCTGTGGGCTCCCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107777 MIM: 600442 MIM: 601383 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AQP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AQP2 - aquaporin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
AQP5 - aquaporin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001651.3 | 982 | Missense Mutation | CGC,TGC | R,C 154 | NP_001642.1 | |
XM_005268838.2 | 982 | Missense Mutation | CGC,TGC | R,C 154 | XP_005268895.1 |
AQP6 - aquaporin 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101927318 - uncharacterized LOC101927318 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |