Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGAATGTGGGGGTGGCACACAG[C/T]GAAGTAAACCCCAACACCCGAGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606092 MIM: 610074 MIM: 610049 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC14 - DnaJ heat shock protein family (Hsp40) member C14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ORMDL2 - ORMDL sphingolipid biosynthesis regulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014182.4 | 116 | Silent Mutation | AGC,AGT | S,S 8 | NP_054901.1 |
SARNP - SAP domain containing ribonucleoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |