Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCGCCCCGCTGGGAGAAGTGAC[C/T]CTCCTGCGCCTGGTGAGTACTTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603826 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAS2L3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAS2L3 - growth arrest specific 2 like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303130.1 | 106 | UTR 5 | NP_001290059.1 | |||
NM_001303131.1 | 106 | Intron | NP_001290060.1 | |||
NM_174942.2 | 106 | UTR 5 | NP_777602.1 | |||
XM_005268804.3 | 106 | UTR 5 | XP_005268861.1 | |||
XM_011538219.2 | 106 | UTR 5 | XP_011536521.1 | |||
XM_011538220.2 | 106 | Intron | XP_011536522.1 | |||
XM_011538221.2 | 106 | Intron | XP_011536523.1 | |||
XM_017019206.1 | 106 | Intron | XP_016874695.1 |
NR1H4 - nuclear receptor subfamily 1 group H member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGAP1 - phosphatidylinositol glycan anchor biosynthesis class A pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |