Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCATATCTCCATCTGCAAATAA[A/T]GGAGATGCCCCAGAAGACCGAGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604979 | ||||||||||||||||||||
Literature Links: |
CPSF6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPSF6 - cleavage and polyadenylation specific factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300947.1 | 239 | Missense Mutation | AAA,AAT | K,N 43 | NP_001287876.1 | |
NM_007007.2 | 239 | Missense Mutation | AAA,AAT | K,N 43 | NP_008938.2 | |
XM_005268588.2 | 239 | Missense Mutation | AAA,AAT | K,N 43 | XP_005268645.1 | |
XM_005268590.2 | 239 | Missense Mutation | AAA,AAT | K,N 43 | XP_005268647.1 |